Gene Transcripts

Gene Symbol
SPRR1A
Entrez Gene ID
6698
Gene Name
small proline rich protein 1A
Gene Aliases
SPRK
Chromosome Location
Chr. 1: 152984088 - 152985814 on Build GRCh38
UniGene ID
Hs.46320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199828.1 NP_001186757.1 2 396
NM_005987.3 NP_005978.2 2 382

View Full Product Details