Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022460.3 | NP_071905.3 | 3 | 264 | |
XM_017004696.1 | XP_016860185.1 | 3 | 267 | ||
XM_017004697.1 | XP_016860186.1 | 3 | 267 | ||
XM_017004698.1 | XP_016860187.1 | 3 | 267 | ||
XM_017004699.1 | XP_016860188.1 | 3 | 319 | ||
XM_017004700.1 | XP_016860189.1 | 3 | 363 | ||
XM_017004701.1 | XP_016860190.1 | 3 | 267 | ||
GenBank mRNA | AK024311.1 | - | 3 | 257 | |
AK314383.1 | - | 3 | 260 | ||
BC050636.1 | - | 3 | 244 | 5193161 | |
BX354596.2 | - | 3 | 295 |