Gene Transcripts
- Gene Symbol
- CDAN1
- Entrez Gene ID
- 146059
- Gene Name
- codanin 1
- Gene Aliases
- CDA1, CDAI, CDAN1A, DLT, PRO1295
- Chromosome Location
- Chr. 15: 42723557 - 42737219 on Build GRCh38
- UniGene ID
- Hs.599232
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138477.2 |
NP_612486.2
|
12, 13 |
1985 |
|
XM_005254176.4 |
XP_005254233.1
|
12, 13 |
1900 |
|
XM_011521270.2 |
XP_011519572.1
|
12, 13 |
1925 |
|
XM_011521271.2 |
XP_011519573.1
|
12, 13 |
1922 |
|
XM_011521274.2 |
XP_011519576.1
|
9, 10 |
971 |
|
GenBank mRNA |
AF525398.1 |
- |
12, 13 |
1982 |
|
BC001092.2 |
- |
4, 5 |
440 |
|
BC008333.1 |
- |
4, 5 |
440 |
|
BC008334.1 |
- |
4, 5 |
440 |
|
BC052568.1 |
- |
12, 13 |
1873 |
|
BC066640.1 |
- |
12, 13 |
1873 |
6577335 |
View Full Product Details