Gene Transcripts

Gene Symbol
GDF6
Entrez Gene ID
392255
Gene Name
growth differentiation factor 6
Gene Aliases
BMP-13, BMP13, CDMP2, KFM, KFS, KFS1, KFSL, LCA17, MCOP4, MCOPCB6, SCDO4, SGM1
Chromosome Location
Chr. 8: 96142330 - 96160792 on Build GRCh38
UniGene ID
Hs.492277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001557.2 NP_001001557.1 2 2193
GenBank mRNA AJ537424.1 - 2 2193

View Full Product Details