Gene Transcripts

Gene Symbol
AMTN
Entrez Gene ID
401138
Gene Name
amelotin
Gene Aliases
UNQ689
Chromosome Location
Chr. 4: 70518572 - 70532743 on Build GRCh38
UniGene ID
Hs.453069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286731.1 NP_001273660.1 3 208
NM_212557.3 NP_997722.1 3 211
GenBank mRNA AB593161.1 - 3 210
AY358528.1 - 3 202
BC121812.1 - 3 179 40109020
BC121817.1 - 3 182 40109016
BF664579.1 - 3 213

View Full Product Details