Gene Transcripts

Gene Symbol
OR4S1
Entrez Gene ID
256148
Gene Name
olfactory receptor family 4 subfamily S member 1
Gene Aliases
OR11-100
Chromosome Location
Chr. 11: 48306223 - 48307152 on Build GRCh38
UniGene ID
Hs.553657
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004725.1 NP_001004725.1 1 902

View Full Product Details