Gene Transcripts
- Gene Symbol
- ASXL2
- Entrez Gene ID
- 55252
- Gene Name
- additional sex combs like 2, transcriptional regulator
- Gene Aliases
- ASXH2
- Chromosome Location
- Chr. 2: 25733758 - 25878485 on Build GRCh38
- UniGene ID
- Hs.119815
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018263.4 |
NP_060733.4
|
10, 11 |
1265 |
|
XM_006712039.3 |
XP_006712102.1
|
6, 7 |
3386 |
|
XM_006712040.2 |
XP_006712103.1
|
6, 7 |
672 |
|
XM_011532950.2 |
XP_011531252.1
|
9, 10 |
1304 |
|
XM_011532951.2 |
XP_011531253.1
|
8, 9 |
992 |
|
XM_017004429.1 |
XP_016859918.1
|
10, 11 |
1095 |
|
XM_017004430.1 |
XP_016859919.1
|
7, 8 |
3452 |
|
GenBank mRNA |
AB051472.2 |
- |
10, 11 |
1265 |
|
AB084279.1 |
- |
10, 11 |
1128 |
|
BC042999.2 |
- |
6, 7 |
703 |
5297392 |
BX647527.1 |
- |
11, 12 |
1338 |
|
BX648218.1 |
- |
10, 11 |
1290 |
|
CR933613.1 |
- |
10, 11 |
1309 |
|
View Full Product Details