Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018211.3 | NP_060681.2 | 3 | 432 | |
XM_006710738.3 | XP_006710801.2 | 3 | 539 | ||
XM_011541706.2 | XP_011540008.1 | 3 | 548 | ||
GenBank mRNA | AB046799.1 | - | 3 | 432 |