Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020348.2 | NP_065081.2 | 11 | 3969 | |
XM_005269692.3 | XP_005269749.2 | 10 | 3819 | ||
XM_011539629.1 | XP_011537931.1 | 12 | 4032 | ||
XM_011539630.1 | XP_011537932.1 | 11 | 3882 | ||
GenBank mRNA | AF169226.1 | - | 11 | 2792 | |
AK056194.1 | - | 11 | 1583 | ||
AL137536.1 | - | 4 | 1338 |