Gene Transcripts

Gene Symbol
OTOP1
Entrez Gene ID
133060
Gene Name
otopetrin 1
Gene Aliases
Chromosome Location
Chr. 4: 4188803 - 4226894 on Build GRCh38
UniGene ID
Hs.534544
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177998.1 NP_819056.1 3 605
GenBank mRNA BC130430.1 - 3 600
BC130432.1 - 3 600 40146463

View Full Product Details