Gene Transcripts

Gene Symbol
Ccdc86
Entrez Gene ID
293738
Gene Name
coiled-coil domain containing 86
Gene Aliases
Chromosome Location
Chr. 1: 226996435 - 227002016 on Build Rnor_6.0
UniGene ID
Rn.40466
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006974.2 NP_001006975.1 1 219
GenBank mRNA BC083770.1 - 1 237
DQ501253.1 - 1 219
FQ213715.1 - 1 258
FQ219684.1 - 1 268

View Full Product Details