Gene Transcripts

Gene Symbol
IRGQ
Entrez Gene ID
126298
Gene Name
immunity related GTPase Q
Gene Aliases
FKSG27, IRGQ1
Chromosome Location
Chr. 19: 43584367 - 43596234 on Build GRCh38
UniGene ID
Hs.6217
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007561.2 NP_001007562.1 3 1214
XM_005258515.3 XP_005258572.1 3 1293
GenBank mRNA AF322648.1 - 2 1244
BC137020.1 - 2 1176 9021013
BC137021.1 - 2 1176 9021014
BC144451.1 - 2 1176 9052977

View Full Product Details