Gene Transcripts

Gene Symbol
Fndc3c1
Entrez Gene ID
333564
Gene Name
fibronectin type III domain containing 3C1
Gene Aliases
Fndc3c, Gm784
Chromosome Location
Chr. X: 106420041 - 106485576 on Build GRCm38
UniGene ID
Mm.298000
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007580.1 NP_001007581.1 15 2440
GenBank mRNA BC076625.1 - 15 2440
BC085176.1 - 15 2438 30626050
DQ192038.1 - 15 2485

View Full Product Details