Gene Transcripts
- Gene Symbol
- Rpl9
- Entrez Gene ID
- 29257
- Gene Name
- ribosomal protein L9
- Gene Aliases
- Chromosome Location
- Chr. 14: 44524419 - 44527613 on Build Rnor_6.0
- UniGene ID
- Rn.107914
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007598.3 |
NP_001007599.3
|
7, 8 |
605 |
|
GenBank mRNA |
BC060589.1 |
- |
7, 8 |
609 |
|
BC086561.1 |
- |
6, 7 |
587 |
7304371 |
FQ210154.1 |
- |
7, 8 |
605 |
|
FQ210235.1 |
- |
7, 8 |
601 |
|
FQ211371.1 |
- |
7, 8 |
605 |
|
FQ212163.1 |
- |
7, 8 |
604 |
|
FQ212174.1 |
- |
7, 8 |
605 |
|
FQ212384.1 |
- |
7, 8 |
609 |
|
FQ212431.1 |
- |
7, 8 |
604 |
|
FQ216865.1 |
- |
7, 8 |
605 |
|
FQ220825.1 |
- |
7, 8 |
603 |
|
FQ221003.1 |
- |
7, 8 |
605 |
|
FQ221028.1 |
- |
7, 8 |
605 |
|
FQ221603.1 |
- |
7, 8 |
605 |
|
FQ221794.1 |
- |
7, 8 |
607 |
|
FQ221999.1 |
- |
7, 8 |
607 |
|
FQ222130.1 |
- |
7, 8 |
606 |
|
FQ222146.1 |
- |
7, 8 |
605 |
|
FQ222204.1 |
- |
7, 8 |
605 |
|
FQ222278.1 |
- |
7, 8 |
603 |
|
FQ222508.1 |
- |
7, 8 |
607 |
|
FQ222509.1 |
- |
7, 8 |
605 |
|
FQ222609.1 |
- |
7, 8 |
607 |
|
FQ222630.1 |
- |
7, 8 |
603 |
|
FQ222743.1 |
- |
7, 8 |
605 |
|
FQ223008.1 |
- |
7, 8 |
619 |
|
FQ223059.1 |
- |
7, 8 |
605 |
|
FQ223395.1 |
- |
7, 8 |
607 |
|
FQ223420.1 |
- |
7, 8 |
603 |
|
FQ223651.1 |
- |
7, 8 |
600 |
|
FQ224555.1 |
- |
7, 8 |
605 |
|
FQ228320.1 |
- |
7, 8 |
606 |
|
FQ228532.1 |
- |
7, 8 |
617 |
|
FQ229128.1 |
- |
7, 8 |
603 |
|
X51706.1 |
- |
6, 7 |
587 |
|
- Gene Symbol
- LOC100360449
- Entrez Gene ID
- 100360449
- Gene Name
- ribosomal protein L9-like
- Gene Aliases
- Chromosome Location
- Chr. 3: 4282206 - 4282911 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100364457
- Entrez Gene ID
- 100364457
- Gene Name
- ribosomal protein L9-like
- Gene Aliases
- Chromosome Location
- Chr. 1: 77829695 - 77830412 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details