Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007622.1 | NP_001007623.1 | 8 | 837 | |
XM_006252255.2 | XP_006252317.1 | 8 | 852 | ||
XM_006252256.2 | XP_006252318.1 | 8 | 847 | ||
XM_006252257.2 | XP_006252319.1 | 8 | 1369 | ||
XM_006252258.1 | XP_006252320.1 | 8 | 737 | ||
XM_006252259.2 | XP_006252321.1 | 8 | 1404 | ||
XM_008770823.1 | XP_008769045.1 | 8 | 738 | ||
GenBank mRNA | AY531526.1 | - | 8 | 819 | |
BC079091.1 | - | 8 | 837 | 7123600 |