Gene Transcripts

Gene Symbol
Thtpa
Entrez Gene ID
305889
Gene Name
thiamine triphosphatase
Gene Aliases
Chromosome Location
Chr. 15: 33753547 - 33759117 on Build Rnor_6.0
UniGene ID
Rn.163276
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007682.1 NP_001007683.1 1 718
XM_006251970.2 XP_006252032.1 1 2649
GenBank mRNA AY065967.1 - 1 80
BC081978.1 - 1 718 7128058

View Full Product Details