Gene Transcripts

Gene Symbol
Rtp2
Entrez Gene ID
224055
Gene Name
receptor transporter protein 2
Gene Aliases
Gm605
Chromosome Location
Chr. 16: 23925548 - 23930847 on Build GRCm38
UniGene ID
Mm.39248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008230.3 NP_001008231.1 2 1948
GenBank mRNA BC049158.1 - 2 1948

View Full Product Details