Gene Transcripts

Gene Symbol
TMEM89
Entrez Gene ID
440955
Gene Name
transmembrane protein 89
Gene Aliases
Chromosome Location
Chr. 3: 48620842 - 48621756 on Build GRCh38
UniGene ID
Hs.631922
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008269.1 NP_001008270.1 2 420
GenBank mRNA BC133034.1 - 2 519
BC133036.1 - 2 519 40147058

View Full Product Details