Gene Transcripts

Gene Symbol
Ltbr
Entrez Gene ID
297604
Gene Name
lymphotoxin beta receptor (TNFR superfamily, member 3)
Gene Aliases
Chromosome Location
Chr. 4: 157822838 - 157829291 on Build Rnor_6.0
UniGene ID
Rn.19329
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008315.1 NP_001008316.1 2 332
XM_006237351.1 XP_006237413.1 2 382
GenBank mRNA BC085880.1 - 2 332

View Full Product Details