Gene Transcripts

Gene Symbol
Rexo2
Entrez Gene ID
300689
Gene Name
RNA exonuclease 2
Gene Aliases
Smfn
Chromosome Location
Chr. 8: 52802604 - 52814321 on Build Rnor_6.0
UniGene ID
Rn.11496
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008326.1 NP_001008327.1 6, 7 743
XM_006243001.2 XP_006243063.1 6, 7 657
GenBank mRNA BC086420.1 - 6, 7 743

View Full Product Details