Gene Transcripts

Gene Symbol
Tom1
Entrez Gene ID
361370
Gene Name
target of myb1 (chicken)
Gene Aliases
Chromosome Location
Chr. 19: 14392367 - 14426482 on Build Rnor_6.0
UniGene ID
Rn.198076
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008365.1 NP_001008366.1 5 428
XM_006255145.2 XP_006255207.1 5 520
GenBank mRNA BC083873.1 - 5 428
FQ213905.1 - 2 100

View Full Product Details