Gene Transcripts

Gene Symbol
Rab1b
Entrez Gene ID
100126191
Gene Name
RAB1B, member RAS oncogene family
Gene Aliases
Rab1bl
Chromosome Location
Chr. 1: 220483529 - 220491469 on Build Rnor_6.0
UniGene ID
Rn.155100
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109979.2 NP_001103449.1 1460
GenBank mRNA BC085118.1 - 1427

Gene Symbol
LOC102555167
Entrez Gene ID
102555167
Gene Name
ras-related protein Rab-1B-like
Gene Aliases
Chromosome Location
Chr. 1: 220480828 - 220482648 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760197.1 XP_008758419.1 1 1423

View Full Product Details