Gene Transcripts

Gene Symbol
Ccl21
Entrez Gene ID
298006
Gene Name
chemokine (C-C motif) ligand 21
Gene Aliases
Ccl21b
Chromosome Location
Chr. 5: 58197678 - 58198782 on Build Rnor_6.0
UniGene ID
Rn.39658
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008513.1 NP_001008513.1 4 535
GenBank mRNA BC086571.1 - 4 535
FQ218040.1 - 4 559
FQ220091.1 - 4 559
FQ225478.1 - 4 559
FQ226079.1 - 4 557
FQ233622.1 - 4 559

View Full Product Details