Gene Transcripts

Gene Symbol
Ndn
Entrez Gene ID
308690
Gene Name
necdin, melanoma antigen (MAGE) family member
Gene Aliases
Chromosome Location
Chr. 1: 122981755 - 122983354 on Build Rnor_6.0
UniGene ID
Rn.3096
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008558.1 NP_001008558.1 1 268
GenBank mRNA BC087630.1 - 1 268

View Full Product Details