Gene Transcripts

Gene Symbol
Krt10
Entrez Gene ID
450225
Gene Name
keratin 10, type I
Gene Aliases
Ka10
Chromosome Location
Chr. 10: 87296445 - 87306614 on Build Rnor_6.0
UniGene ID
Rn.125065
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008804.1 NP_001008804.1 1 542
XM_006247391.2 XP_006247453.1 1 6439
XM_008768091.1 XP_008766313.1 1 6439

View Full Product Details