Gene Transcripts

Gene Symbol
Vom1r7
Entrez Gene ID
494259
Gene Name
vomeronasal 1 receptor 7
Gene Aliases
V1re3
Chromosome Location
Chr. 1: 60087495 - 60088406 on Build Rnor_6.0
UniGene ID
Rn.129458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008907.1 NP_001008907.1 1 822
GenBank mRNA AY510304.1 - 1 822

View Full Product Details