Gene Transcripts

Gene Symbol
Vom1r61
Entrez Gene ID
494282
Gene Name
vomeronasal 1 receptor 61
Gene Aliases
V1rj3
Chromosome Location
Chr. 1: 75298364 - 75299317 on Build Rnor_6.0
UniGene ID
Rn.129408
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008935.1 NP_001008935.1 1 161
GenBank mRNA AY510336.1 - 1 161

View Full Product Details