Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009539.1 | NP_001009539.1 | 8 | 1664 | |
XM_006250253.2 | XP_006250315.1 | 8 | 1695 | ||
XM_006250254.2 | XP_006250316.1 | 8 | 1815 | ||
GenBank mRNA | BC061812.1 | - | 8 | 1664 |