Gene Transcripts
- Gene Symbol
- ODF2L
- Entrez Gene ID
- 57489
- Gene Name
- outer dense fiber of sperm tails 2 like
- Gene Aliases
- dJ977L11.1
- Chromosome Location
- Chr. 1: 86346824 - 86396345 on Build GRCh38
- UniGene ID
- Hs.149360
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007022.2 |
NP_001007023.2
|
7, 8 |
996 |
|
NM_001184765.1 |
NP_001171694.1
|
7, 8 |
1003 |
|
NM_001184766.1 |
NP_001171695.1
|
7, 8 |
1003 |
|
NM_020729.2 |
NP_065780.2
|
7, 8 |
1003 |
|
XM_005271054.3 |
XP_005271111.1
|
7, 8 |
1001 |
|
XM_011541815.2 |
XP_011540117.1
|
7, 8 |
1001 |
|
XM_011541819.2 |
XP_011540121.1
|
7, 8 |
1001 |
|
XM_011541821.2 |
XP_011540123.1
|
7, 8 |
1001 |
|
XM_011541823.2 |
XP_011540125.1
|
7, 8 |
1001 |
|
XM_011541824.2 |
XP_011540126.1
|
7, 8 |
1001 |
|
XM_017001876.1 |
XP_016857365.1
|
7, 8 |
793 |
|
XM_017001877.1 |
XP_016857366.1
|
7, 8 |
793 |
|
XM_017001878.1 |
XP_016857367.1
|
6, 7 |
884 |
|
XM_017001879.1 |
XP_016857368.1
|
7, 8 |
1001 |
|
XM_017001880.1 |
XP_016857369.1
|
6, 7 |
884 |
|
XM_017001881.1 |
XP_016857370.1
|
5, 6 |
756 |
|
GenBank mRNA |
AB033055.1 |
- |
7, 8 |
916 |
|
AK302792.1 |
- |
7, 8 |
975 |
|
AK303209.1 |
- |
7, 8 |
1000 |
|
AK308351.1 |
- |
4, 5 |
526 |
|
BC009779.1 |
- |
3, 4 |
400 |
|
BC029420.1 |
- |
7, 8 |
721 |
|
BC047368.1 |
- |
7, 8 |
943 |
5276959 |
BC091490.1 |
- |
7, 8 |
902 |
30367572 |
BX499763.1 |
- |
3, 4 |
217 |
|
View Full Product Details