Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001284355.2 | NP_001271284.1 | 6 | 4019 | |
NM_133463.3 | NP_597720.1 | 7 | 4244 | ||
XM_011515150.2 | XP_011513452.1 | 7 | 4189 | ||
GenBank mRNA | AB075830.1 | - | 7 | 4237 | |
BC146753.1 | - | 7 | 4237 | 9007189 |