Gene Transcripts

Gene Symbol
Sgce
Entrez Gene ID
432360
Gene Name
sarcoglycan, epsilon
Gene Aliases
Chromosome Location
Chr. 4: 29726140 - 29769902 on Build Rnor_6.0
UniGene ID
Rn.185815
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002023.2 NP_001002023.1 6 890
GenBank mRNA AY164274.1 - 6 851
BC089947.1 - 6 890 7311584

View Full Product Details