Gene Transcripts

Gene Symbol
Npc1l1
Entrez Gene ID
432367
Gene Name
NPC1-like 1
Gene Aliases
Chromosome Location
Chr. 14: 86345946 - 86560694 on Build Rnor_6.0
UniGene ID
Rn.123563
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002025.1 NP_001002025.1 4 1726
GenBank mRNA AY437867.1 - 4 1726

View Full Product Details