Gene Transcripts

Gene Symbol
Rab44
Entrez Gene ID
442827
Gene Name
RAB44, member RAS oncogene family
Gene Aliases
9830134C10Rik
Chromosome Location
Chr. 17: 29114128 - 29148980 on Build GRCm38
UniGene ID
Mm.440341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002786.2 NP_001002786.1 8 3547
GenBank mRNA AK036563.1 - 8 3548

View Full Product Details