Gene Transcripts

Gene Symbol
Dnmt3l
Entrez Gene ID
309680
Gene Name
DNA (cytosine-5-)-methyltransferase 3-like
Gene Aliases
Chromosome Location
Chr. 20: 11344513 - 11359090 on Build Rnor_6.0
UniGene ID
Rn.129905
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003964.1 NP_001003964.1 2 180
XM_006256250.2 XP_006256312.1 1 203

View Full Product Details