Gene Transcripts

Gene Symbol
Txndc8
Entrez Gene ID
362525
Gene Name
thioredoxin domain containing 8
Gene Aliases
Sptrx3
Chromosome Location
Chr. 5: 75088071 - 75116427 on Build Rnor_6.0
UniGene ID
Rn.67443
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004092.2 NP_001004092.1 1, 2 57
GenBank mRNA AY496270.1 - 1, 2 60

View Full Product Details