Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014797.2 | NP_055612.2 | 7 | 2457 | |
GenBank mRNA | AB007901.1 | - | 7 | 2457 | |
BC117374.1 | - | 6 | 2325 | 40125925 | |
BC117376.1 | - | 6 | 2325 | 40125927 | |
BX648883.1 | - | 1633 |