Gene Transcripts

Gene Symbol
Emc10
Entrez Gene ID
292878
Gene Name
ER membrane protein complex subunit 10
Gene Aliases
Inm02
Chromosome Location
Chr. 1: 100495115 - 100501453 on Build Rnor_6.0
UniGene ID
Rn.1470
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004221.1 NP_001004221.1 7 1502
XM_006229000.2 XP_006229062.1 8 1605
GenBank mRNA BC079041.1 - 7 1502

View Full Product Details