Gene Transcripts

Gene Symbol
OR52N4
Entrez Gene ID
390072
Gene Name
olfactory receptor family 52 subfamily N member 4 (gene/pseudogene)
Gene Aliases
OR11-64
Chromosome Location
Chr. 11: 5722137 - 5755958 on Build GRCh38
UniGene ID
Hs.553735
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005175.3 NP_001005175.3 1 979
XM_017017711.1 XP_016873200.1 5 5527
XM_017017712.1 XP_016873201.1 7 5687
XM_017017713.1 XP_016873202.1 4 5496
GenBank mRNA BC137035.1 - 1 979
BC137036.1 - 1 979 9021032

View Full Product Details