Gene Transcripts

Gene Symbol
Zcwpw1
Entrez Gene ID
381678
Gene Name
zinc finger, CW type with PWWP domain 1
Gene Aliases
Gm1053
Chromosome Location
Chr. 5: 137787395 - 137822621 on Build GRCm38
UniGene ID
Mm.332765
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005426.2 NP_001005426.2 16, 17 1751
GenBank mRNA BC071186.1 - 16, 17 1678
CF982070.1 - 6, 7 472

View Full Product Details