Gene Transcripts

Gene Symbol
Slc16a13
Entrez Gene ID
287451
Gene Name
solute carrier family 16, member 13
Gene Aliases
Chromosome Location
Chr. 10: 56827593 - 56831243 on Build Rnor_6.0
UniGene ID
Rn.23260
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005530.1 NP_001005530.1 4 1405
GenBank mRNA BC081902.1 - 4 1405

View Full Product Details