Gene Transcripts

Gene Symbol
Repin1
Entrez Gene ID
445541
Gene Name
replication initiator 1
Gene Aliases
Ap-4, Ap4
Chromosome Location
Chr. 4: 78231772 - 78237498 on Build Rnor_6.0
UniGene ID
Rn.204397
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005893.1 NP_001005893.1 1 140
XM_006236423.2 XP_006236485.1 2 1039
XM_006236424.2 XP_006236486.1 2 1036
XM_006236425.2 XP_006236487.1 3 1091
XM_006236426.2 XP_006236488.1 3 1089
XM_006236427.2 XP_006236489.1 3 1160
XM_006236428.2 XP_006236490.1 2 963
XM_006236429.2 XP_006236491.1 2 961
GenBank mRNA AY691175.1 - 1 140

View Full Product Details