Gene Transcripts
- Gene Symbol
- TECPR2
- Entrez Gene ID
- 9895
- Gene Name
- tectonin beta-propeller repeat containing 2
- Gene Aliases
- KIAA0329, SPG49
- Chromosome Location
- Chr. 14: 102362912 - 102502481 on Build GRCh38
- UniGene ID
- Hs.195667
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172631.2 |
NP_001166102.1
|
2, 3 |
488 |
|
NM_014844.4 |
NP_055659.2
|
2, 3 |
488 |
|
GenBank mRNA |
AB002327.1 |
- |
2, 3 |
437 |
|
AK127721.1 |
- |
2, 3 |
921 |
|
AK296395.1 |
- |
2, 3 |
339 |
|
BC136647.1 |
- |
1, 2 |
285 |
9020637 |
BC142667.1 |
- |
2, 3 |
321 |
40148157 |
BC142715.1 |
- |
2, 3 |
321 |
40148843 |
BC151230.1 |
- |
2, 3 |
461 |
8860116 |
BP365224.1 |
- |
2, 3 |
490 |
|
View Full Product Details