Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145123.2 | NP_001138595.1 | 4 | 1126 | |
NM_006658.4 | NP_006649.2 | 5 | 1279 | ||
XM_011515094.1 | XP_011513396.1 | 6 | 911 | ||
GenBank mRNA | AF071789.1 | - | 4 | 1004 | |
AF097730.1 | - | 4 | 1001 | ||
AK293478.1 | - | 4 | 928 | ||
BC028094.1 | - | 5 | 1063 | 5241722 |