Gene Transcripts

Gene Symbol
Sucnr1
Entrez Gene ID
408199
Gene Name
succinate receptor 1
Gene Aliases
Gpr91
Chromosome Location
Chr. 2: 150211897 - 150212848 on Build Rnor_6.0
UniGene ID
Rn.124512
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001518.1 NP_001001518.1 1 508
GenBank mRNA AY612851.1 - 1 508

View Full Product Details