Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242925.1 | NP_001229854.1 | 3 | 1636 | |
NM_001297611.1 | NP_001284540.1 | 2 | 1094 | ||
NM_001297613.1 | NP_001284542.1 | 2 | 1114 | ||
NM_014858.3 | NP_055673.2 | 3 | 2300 | ||
XM_005245684.1 | XP_005245741.1 | 3 | 1991 | ||
XM_005245685.4 | XP_005245742.1 | 3 | 1622 | ||
XM_005245686.3 | XP_005245743.1 | 3 | 2017 | ||
XM_017003000.1 | XP_016858489.1 | 2 | 1075 | ||
GenBank mRNA | AB001596.1 | - | 2 | 970 | |
AB007950.3 | - | 3 | 2300 | ||
AK095816.1 | - | 2 | 1114 | ||
AK131419.1 | - | 4 | 2089 | ||
AK293734.1 | - | 3 | 1636 | ||
AK314398.1 | - | 2 | 993 | ||
AY358192.1 | - | 2 | 2208 | ||
BC041019.1 | - | 2 | 1034 | ||
BC053876.1 | - | 758 | |||
BC131629.1 | - | 3 | 1772 | 40118729 |