Gene Transcripts

Gene Symbol
Oxgr1
Entrez Gene ID
239283
Gene Name
oxoglutarate (alpha-ketoglutarate) receptor 1
Gene Aliases
Cysltr3, Gm299, Gpr80, Gpr99, P2Y15
Chromosome Location
Chr. 14: 120019585 - 120042435 on Build GRCm38
UniGene ID
Mm.138520
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001490.3 NP_001001490.1 4 609
GenBank mRNA AK142510.1 - 4 610
AY612852.1 - 1 86
BC119196.1 - 1 139 8733945
BC119200.1 - 1 139 8733949

View Full Product Details