Gene Transcripts

Gene Symbol
Mrgprb3
Entrez Gene ID
404238
Gene Name
MAS-related GPR, member B3
Gene Aliases
MrgB3
Chromosome Location
Chr. 7: 48642863 - 48643801 on Build GRCm38
UniGene ID
Mm.377926
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207537.1 NP_997420.1 1 34
GenBank mRNA BC107192.1 - 1 33
BC107193.1 - 1 33
BC114400.1 - 1 33
BC114428.1 - 1 33

View Full Product Details