Gene Transcripts
- Gene Symbol
- Cdk10
- Entrez Gene ID
- 234854
- Gene Name
- cyclin-dependent kinase 10
- Gene Aliases
- BC017131
- Chromosome Location
- Chr. 8: 123224840 - 123232256 on Build GRCm38
- UniGene ID
- Mm.190984
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_194444.2 |
NP_919426.2
|
2, 3 |
157 |
|
NM_194446.2 |
NP_919428.1
|
2, 3 |
195 |
|
GenBank mRNA |
AK133918.1 |
- |
2, 3 |
150 |
|
AK144801.1 |
- |
2, 3 |
167 |
|
AK170594.1 |
- |
2, 3 |
167 |
|
BC017131.1 |
- |
2, 3 |
132 |
4506458 |
BC023736.1 |
- |
2, 3 |
176 |
|
BC052096.1 |
- |
2, 3 |
164 |
|
BY010457.1 |
- |
2, 3 |
158 |
|
CJ102037.1 |
- |
2, 3 |
157 |
|
DQ288857.1 |
- |
2, 3 |
194 |
|
View Full Product Details