Gene Transcripts

Gene Symbol
SPG20
Entrez Gene ID
23111
Gene Name
spastic paraplegia 20 (Troyer syndrome)
Gene Aliases
SPARTIN, TAHCCP1
Chromosome Location
Chr. 13: 36301638 - 36370180 on Build GRCh38
UniGene ID
Hs.440414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142294.1 NP_001135766.1 2 247
NM_001142295.1 NP_001135767.1 2 371
NM_001142296.1 NP_001135768.1 3 423
NM_015087.4 NP_055902.1 2 440
XM_005266313.4 XP_005266370.1 2 365
XM_005266314.3 XP_005266371.1 3 414
XM_005266315.3 XP_005266372.1 3 268
XM_005266317.3 XP_005266374.1 2 198
XM_011535012.2 XP_011533314.1 2 351
GenBank mRNA AB011182.1 - 2 214
AK001949.1 - 2 204
AK057764.1 - 2 220
AK291724.1 - 2 291
AY038359.1 - 1 153
AY123329.1 - 2 214
BC047083.1 - 2 247 5313378
BX648004.1 - 2 336
DA347686.1 - 2 328
DA507821.1 - 2 351
DA710623.1 - 2 415
KU178563.1 - 1 153
KU178564.1 - 1 153

View Full Product Details