Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014939.3 | NP_055754.2 | 23 | 3849 | |
XM_005258233.2 | XP_005258290.1 | 23 | 3942 | ||
XM_005258234.2 | XP_005258291.1 | 23 | 3942 | ||
XM_005258235.3 | XP_005258292.1 | 18 | 2723 | ||
XM_006722420.3 | XP_006722483.1 | 23 | 3942 | ||
XM_011525884.2 | XP_011524186.1 | 23 | 3942 | ||
XM_017025615.1 | XP_016881104.1 | 23 | 3441 | ||
XM_017025616.1 | XP_016881105.1 | 23 | 3939 | ||
XM_017025617.1 | XP_016881106.1 | 23 | 3939 | ||
GenBank mRNA | AB023229.1 | - | 23 | 3542 | |
AK001662.1 | - | 15 | 2208 | ||
AK292537.1 | - | 23 | 3828 | ||
AL136749.1 | - | 24 | 3374 | ||
BC127109.1 | - | 23 | 3403 | 40124744 | |
BC127110.1 | - | 23 | 3403 | 40124745 |